Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-Amino-2-piperidone 98.0+%, TCI America™
SDP

Catalog No. A31715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31715G 5 g
A31711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31715G Supplier TCI America Supplier No. A31715G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34294-79-6
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.15
MDL Number MFCD09258624
InChI Key YCCMTCQQDULIFE-UHFFFAOYNA-N
PubChem CID 7200369
IUPAC Name 3-aminopiperidin-2-one
SMILES NC1CCCNC1=O

Specifications

Melting Point 87°C
Color Yellow
Quantity 5 g
Formula Weight 114.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-3-Amino-2-piperidone
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.