Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic Acid 95.0+%, TCI America™
SDP

Catalog No. A24701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24701G 1 g
A24705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24701G Supplier TCI America Supplier No. A24701G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73259-81-1
Molecular Formula C8H16N2O4
Molecular Weight (g/mol) 204.23
MDL Number MFCD00236843
InChI Key KRJLRVZLNABMAT-YFKPBYRVSA-N
Synonym boc-dap-oh, 3-amino-boc-l-alanine, boc-l-2,3-diaminopropionic acid, s-3-amino-2-tert-butoxycarbonyl amino propanoic acid, n-boc-beta-amino-ala-oh, boc-dpr-oh, 2s-3-amino-2-tert-butoxy carbonyl amino propanoic acid, 3-amino-n-boc-l-alanine, n-boc-l-2,3-diaminopropanoic acid, nalpha-boc-2,3-diaminopropionic acid
PubChem CID 2755946
IUPAC Name (2S)-3-azaniumyl-2-{[(tert-butoxy)carbonyl]amino}propanoate
SMILES CC(C)(C)OC(=O)N[C@@H](C[NH3+])C([O-])=O

Specifications

Melting Point 210°C
Color White
Quantity 1 g
Formula Weight 204.23
Percent Purity ≥95.0% (HPLC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-Amino-2-(tert-butoxycarbonylamino)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.