Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-3-amino-4-hydroxybutanoic acid | 16504-57-7 | MFCD00270241 | ≥97.0% | 119.12 | C4H9NO3 | 10 G
SDP

Catalog No. 5000209344
Encompass_Preferred

(S)-3-amino-4-hydroxybutanoic acid is the S-enantiomer of 3-amino-4-hydroxybutanoic acid, a serine-derived amino acid derivative provided for research use. It is commonly used as a chiral building block or reference in biochemical and neuropharmacological studies.

  • Chiral S-enantiomer suitable for enantioselective studies.
  • High purity: ≥97.0%.
  • Molecular weight 119.12 g·mol⁻¹.
  • Chemical formula C4H9NO3.
  • Available as a 10 G quantity for laboratory use.
  • Powder storage recommended at -20°C (long term) or 4°C (intermediate term).

Catalog No. 50-002-09344 Supplier Medchemexpress LLC Supplier No. HYW04043210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-3-amino-4-hydroxybutanoic acid is the S-enantiomer of 3-amino-4-hydroxybutanoic acid, a serine-derived amino acid derivative provided for research use. It is commonly used as a chiral building block or reference in biochemical and neuropharmacological studies.

  • Chiral S-enantiomer suitable for enantioselective studies.
  • High purity: ≥97.0%.
  • Molecular weight 119.12 g·mol⁻¹.
  • Chemical formula C4H9NO3.
  • Available as a 10 G quantity for laboratory use.
  • Powder storage recommended at -20°C (long term) or 4°C (intermediate term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.