Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-3-(aminomethyl)-4-chloro-7-(2-hydroxyethoxy)benzo[c]oxaborol-1(3H)-ol | 2131798-13-3 | MFCD31657361 | 100.0% | 293.94 g/mol | C10H14BCl2NO4 | 1 ML
SDP

Catalog No. 5000240899
Encompass_Preferred

Ganfeborole hydrochloride is a boron-containing small-molecule antitubercular agent that inhibits Mycobacterium tuberculosis leucyl-tRNA synthetase (LeuRS) with an IC50 of 0.2 μM. It is supplied as a high-purity research compound in solution or solid form for biochemical and microbiology studies.

  • Inhibits M. tuberculosis LeuRS with an IC50 of 0.2 μM.
  • High purity suitable for research applications.
  • Supplied as a 10 mM solution in DMSO; solid formats available.
  • Soluble in DMSO (≈67.5 mg/mL); ultrasonic agitation recommended.
  • Store sealed and protected from moisture; long-term solution storage at -80°C.
  • Molecular weight 293.94 g/mol; formula C10H14BCl2NO4.

Catalog No. 50-002-40899 Supplier Medchemexpress LLC Supplier No. HY10777510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ganfeborole hydrochloride is a boron-containing small-molecule antitubercular agent that inhibits Mycobacterium tuberculosis leucyl-tRNA synthetase (LeuRS) with an IC50 of 0.2 μM. It is supplied as a high-purity research compound in solution or solid form for biochemical and microbiology studies.

  • Inhibits M. tuberculosis LeuRS with an IC50 of 0.2 μM.
  • High purity suitable for research applications.
  • Supplied as a 10 mM solution in DMSO; solid formats available.
  • Soluble in DMSO (≈67.5 mg/mL); ultrasonic agitation recommended.
  • Store sealed and protected from moisture; long-term solution storage at -80°C.
  • Molecular weight 293.94 g/mol; formula C10H14BCl2NO4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.