Learn More
Medchemexpress LLC (S)-3-(aminomethyl)-dioxolo[4',5':5,6]benzo[1,2-c]oxaborol-1(3H)-ol hydrochloride | 1853176-89-2 | 99.3% | 243.45 g/mol | C9H11BClNO4 | 100MG
SDP
Catalog No. 5000236319
Encompass_Preferred
DS86760016 is a leucyl-tRNA synthetase (LeuRS) inhibitor intended for antibacterial research; it is provided as a hydrochloride salt and has reported activity against multidrug-resistant Gram-negative bacteria. Reported enzyme IC50 values include Escherichia coli 0.38 μM, Pseudomonas aeruginosa 0.62 μM, and Acinetobacter baumannii 0.16 μM.
  • Potent LeuRS inhibition with low micromolar IC50s (E. coli 0.38 μM; P. aeruginosa 0.62 μM; A. baumannii 0.16 μM).
  • High reported purity (99.3%).
  • Characterized chemical identity: C9H11BClNO4, MW 243.45 g/mol.
  • Available in multiple pack sizes including 100 mg for laboratory use.
  • Suitable for in vitro antibacterial assays and preclinical research.
Catalog No. 50-002-36319 Supplier Medchemexpress LLC Supplier No. HY124679100MG
DS86760016 is a leucyl-tRNA synthetase (LeuRS) inhibitor intended for antibacterial research; it is provided as a hydrochloride salt and has reported activity against multidrug-resistant Gram-negative bacteria. Reported enzyme IC50 values include Escherichia coli 0.38 μM, Pseudomonas aeruginosa 0.62 μM, and Acinetobacter baumannii 0.16 μM.
  • Potent LeuRS inhibition with low micromolar IC50s (E. coli 0.38 μM; P. aeruginosa 0.62 μM; A. baumannii 0.16 μM).
  • High reported purity (99.3%).
  • Characterized chemical identity: C9H11BClNO4, MW 243.45 g/mol.
  • Available in multiple pack sizes including 100 mg for laboratory use.
  • Suitable for in vitro antibacterial assays and preclinical research.