Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-Aminoquinuclidine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. A24341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24341G Supplier TCI America Supplier No. A24341G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 119904-90-4
Molecular Formula C7H16Cl2N2
Molecular Weight (g/mol) 199.119
MDL Number MFCD00191753
InChI Key STZHBULOYDCZET-XCUBXKJBSA-N
Synonym (S)-(-)-3-Amino-1-azabicyclo[2.2.2]octane Dihydrochloride
PubChem CID 12494973
IUPAC Name (3S)-1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
SMILES C1CN2CCC1C(C2)N.Cl.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 199.12
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-3-Aminoquinuclidine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.