Learn More
Medchemexpress LLC (S)-3-(azetidin-2-ylmethoxy)-5-iodopyridine dihydrochloride | 1217837-17-6 | 99.0% | 363.02 | C9H13Cl2IN2O | 10 MG
SDP
Catalog No. 5000242097
Encompass_Preferred
5-Iodo-A-85380 dihydrochloride is an iodinated, high-affinity ligand selective for neuronal nicotinic acetylcholine receptors (nAChRs), with potent binding at the α4β2 subtype. It is supplied for research applications in receptor binding and pharmacology, and has reported Kd values of ~12 pM (rat) and ~14 pM (human).
  • High affinity for α4β2 nAChR enabling sensitive binding assays.
  • Demonstrated selectivity for neuronal nicotinic receptor subtypes.
  • High reported purity for reliable experimental results.
  • Solid form with defined storage recommendations to preserve stability.
  • Suitable for in vitro pharmacology and receptor characterization studies.
Catalog No. 50-002-42097 Supplier Medchemexpress LLC Supplier No. HY10768610MG
5-Iodo-A-85380 dihydrochloride is an iodinated, high-affinity ligand selective for neuronal nicotinic acetylcholine receptors (nAChRs), with potent binding at the α4β2 subtype. It is supplied for research applications in receptor binding and pharmacology, and has reported Kd values of ~12 pM (rat) and ~14 pM (human).
  • High affinity for α4β2 nAChR enabling sensitive binding assays.
  • Demonstrated selectivity for neuronal nicotinic receptor subtypes.
  • High reported purity for reliable experimental results.
  • Solid form with defined storage recommendations to preserve stability.
  • Suitable for in vitro pharmacology and receptor characterization studies.