Learn More
Medchemexpress LLC (S)-3-(azetidin-2-ylmethoxy)-5-iodopyridine dihydrochloride | 1217837-17-6 | MFCD03791315 | 99.0% | 363.02 g·mol⁻1 | C9H13Cl2IN2O | 25 MG
SDP
Catalog No. 5000242189
Encompass_Preferred
5-Iodo-A-85380 dihydrochloride is a high-affinity, small-molecule ligand and agonist for neuronal nicotinic acetylcholine receptors, with strong selectivity for the α4β2 subtype. It is supplied as the dihydrochloride salt for research use only and has a reported binding Kd of ≈12-14 pM.
  • High affinity for α4β2 nicotinic acetylcholine receptors (Kd ≈12-14 pM).
  • Selective agonist activity at α4β2 and α6β2 receptor subtypes.
  • Supplied as the dihydrochloride salt for improved stability and handling.
  • High purity suitable for research use (≈99.0%).
  • Available in small-scale pack sizes for laboratory studies.
Catalog No. 50-002-42189 Supplier Medchemexpress LLC Supplier No. HY10768625MG
5-Iodo-A-85380 dihydrochloride is a high-affinity, small-molecule ligand and agonist for neuronal nicotinic acetylcholine receptors, with strong selectivity for the α4β2 subtype. It is supplied as the dihydrochloride salt for research use only and has a reported binding Kd of ≈12-14 pM.
  • High affinity for α4β2 nicotinic acetylcholine receptors (Kd ≈12-14 pM).
  • Selective agonist activity at α4β2 and α6β2 receptor subtypes.
  • Supplied as the dihydrochloride salt for improved stability and handling.
  • High purity suitable for research use (≈99.0%).
  • Available in small-scale pack sizes for laboratory studies.