Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid, 98%, Thermo Scientific™

Catalog No. AAH2705606
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2705606 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2705606 Supplier Thermo Scientific Chemicals Supplier No. H2705606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 97534-82-2
Molecular Formula C12H13NO5
Quantity 5g
MDL Number MFCD00274193
Synonym s-3-benzyloxy carbonyl oxazolidine-4-carboxylic acid, s---3-benzyloxycarbonyl-4-oxazolidinecarboxylic acid, s---3-z-4-oxazolidinecarboxylic acid, s 3-benzyloxycarbonyl oxazolidine-4-carboxylic acid, 4s-3-benzyloxy carbonyl-1,3-oxazolidine-4-carboxylic acid, s-cbz-oxaproline, s---3-z-4-oxazolidinecarboxylicacid, 4s-3-benzyloxycarbonyloxazolidine-4-carboxylic acid, s-+-3-cbz-4-oxazolidinecarboxylic acid, s-oxazolidine-3,4-dicarboxylic acid 3-benzyl ester
Optical Rotation −79° (c=1 in Chloroform)
InChI Key XRRRGBIMHQARMF-JTQLQIEISA-N
SMILES C1C(N(CO1)C(=O)OCC2=CC=CC=C2)C(=O)O
IUPAC Name (4S)-3-phenylmethoxycarbonyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol) 251.238
PubChem CID 688328
Formula Weight 251.24
Percent Purity 98%
Chemical Name or Material (S)-(-)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.