Learn More
Medchemexpress LLC (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde | 102308-32-7 | 97.9% | C11H19NO4 | 25 G
SDP
Catalog No. 5000359173
Encompass_Preferred
(S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is a chiral compound commonly utilized as a chiral intermediate in asymmetric synthesis. It holds significant applications in organic synthesis and can enhance the polarity, solubility, and the potential for forming hydrogen bonds in compounds as a polar aldehyde substrate.
  • Chiral compound
  • Commonly used as a chiral intermediate in asymmetric synthesis
  • Has important applications in the field of organic synthesis
  • Can enhance the polarity, solubility, and the possibility of forming hydrogen bonds of compounds
Catalog No. 50-003-59173 Supplier Medchemexpress LLC Supplier No. 25GHY69348
(S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is a chiral compound commonly utilized as a chiral intermediate in asymmetric synthesis. It holds significant applications in organic synthesis and can enhance the polarity, solubility, and the potential for forming hydrogen bonds in compounds as a polar aldehyde substrate.
  • Chiral compound
  • Commonly used as a chiral intermediate in asymmetric synthesis
  • Has important applications in the field of organic synthesis
  • Can enhance the polarity, solubility, and the possibility of forming hydrogen bonds of compounds