Learn More
Medchemexpress LLC (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde | 102308-32-7 | 97.9% | C11H19NO4 | 5 G
SDP
Catalog No. 5000434234
Encompass_Preferred
(S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is a chiral compound commonly used as an intermediate in asymmetric synthesis. It has important applications in organic synthesis and, as a polar aldehyde substrate, can enhance the polarity, solubility, and hydrogen-bonding potential of compounds.
  • Commonly used as a chiral intermediate in asymmetric synthesis
  • Important applications in organic synthesis
  • As a polar aldehyde substrate, it can enhance polarity, solubility, and the possibility of forming hydrogen bonds of compounds
Catalog No. 50-004-34234 Supplier Medchemexpress LLC Supplier No. HY693485G
(S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is a chiral compound commonly used as an intermediate in asymmetric synthesis. It has important applications in organic synthesis and, as a polar aldehyde substrate, can enhance the polarity, solubility, and hydrogen-bonding potential of compounds.
  • Commonly used as a chiral intermediate in asymmetric synthesis
  • Important applications in organic synthesis
  • As a polar aldehyde substrate, it can enhance polarity, solubility, and the possibility of forming hydrogen bonds of compounds