Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde, 95%, Thermo Scientific™

Catalog No. AAH51049MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51049MD 250 mg
AAH5104903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51049MD Supplier Thermo Scientific Chemicals Supplier No. H51049MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used to prepare γ-aminophosphonates (N-arylamide phosphonates as subtype-selective sphingosine 1-phosphate receptor agonists and antagonists. It is also used to synthesize N-sulfonylated propargylglycine derivatives as metalloproteinase inhibitors. Also widely utilized in the asymmetric synthesis of natural products.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102308-32-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00209557
InChI Key PNJXYVJNOCLJLJ-MRVPVSSYSA-N
Synonym s---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl s---4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, tert-butyl 4s-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, unii-41aj913hqi, s-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, 1,1-dimethylethyl-s-4-formyl-2,2-dimethyl-3-oxazolidine carboxylate, s---3-tert-butoxycarbonyl-4-formyl-2,2-dimethyl-1,3-oxazolidine, s-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4s, s---3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine
PubChem CID 179824
IUPAC Name tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
SMILES CC1(N(C(CO1)C=O)C(=O)OC(C)(C)C)C

Specifications

Quantity 250 mg
Beilstein 3591324
Merck Index 14,4371
Optical Rotation −90° (c=1 in Chloroform)
Formula Weight 229.27
Percent Purity 95%
Chemical Name or Material (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.