Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-(Boc-amino)-4-(4-pyridyl)butyric acid, 95%, Thermo Scientific™

Catalog No. AAH52197MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-3-(Boc-amino)-4-(4-pyridyl)butyric acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 219297-13-9
Molecular Formula C14H20N2O4
MDL Number MFCD01076288
Synonym s-3-tert-butoxycarbonyl amino-4-pyridin-4-yl butanoic acid, s-3-boc-amino-4-4-pyridyl butyric acid, 3s-3-tert-butoxycarbonyl amino-4-pyridin-4-yl butanoic acid, s-boc-4-4-pyridyl-beta-homoala-oh, s-boc-4-4-pyridyl-beta-homoala-oh hplc, 3s-3-2-methylpropan-2-yl oxycarbonylamino-4-pyridin-4-ylbutanoic acid
InChI Key BYTKQINZDKLYGV-NSHDSACASA-N
SMILES CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)CC(=O)O
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-pyridin-4-ylbutanoic acid
Molecular Weight (g/mol) 280.324
PubChem CID 2761651
Formula Weight 280.32
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-4-(4-pyridyl)butyric acid
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.