Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-3-(Boc-amino)-4-phenylbutyric acid | 51871-62-6 | MFCD01076271 | ≥95.0% | 279.33 g/mol | C15H21NO4 | 1 G
SDP

Catalog No. 5000210460
Encompass_Preferred

(S)-3-(Boc-amino)-4-phenylbutyric acid is a Boc-protected phenylalanine derivative used as a research reagent and synthetic intermediate. It is supplied as a white to off-white solid and is commonly used in peptide synthesis and medicinal chemistry.

  • Appearance: White to off-white solid.
  • CAS number: 51871-62-6.
  • Molecular formula: C15H21NO4.
  • Molecular weight: 279.33 g/mol.
  • Purity: ≥95.0%.
  • Available pack sizes: 250 mg, 1 g, 5 g, 10 g, 25 g.
  • Storage: Powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-10460 Supplier Medchemexpress LLC Supplier No. HYW0424781G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-3-(Boc-amino)-4-phenylbutyric acid is a Boc-protected phenylalanine derivative used as a research reagent and synthetic intermediate. It is supplied as a white to off-white solid and is commonly used in peptide synthesis and medicinal chemistry.

  • Appearance: White to off-white solid.
  • CAS number: 51871-62-6.
  • Molecular formula: C15H21NO4.
  • Molecular weight: 279.33 g/mol.
  • Purity: ≥95.0%.
  • Available pack sizes: 250 mg, 1 g, 5 g, 10 g, 25 g.
  • Storage: Powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.