Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-(Boc-amino)-5-phenylpentanoic acid, 95%, Thermo Scientific Chemicals

Catalog No. aah5218003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5218003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5218003 Supplier Thermo Scientific Chemicals Supplier No. H5218003
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 218608-84-5
Molecular Formula C16H23NO4
Molecular Weight (g/mol) 293.36
MDL Number MFCD01076264,MFCD05863635,MFCD01076262
InChI Key MYWZFJXOLAXENE-UHFFFAOYNA-N
Synonym r-3-tert-butoxycarbonyl amino-5-phenylpentanoic acid, r-3-boc-amino-5-phenylpentanoic acid, 3r-3-tert-butoxycarbonyl amino-5-phenylpentanoic acid, boc-d-?-nva 5-phenyl-oh, n-boc-3-r-amino-5-phenylpentanoic acid, n-tert-butoxycarbonyl-3 r-amino-5-phenylpentanoic acid, 3r-3-tert-butoxycarbonylamino-5-phenyl-pentanoic acid, r-3-tert-butoxycarbonyl amino-5-phenylpentanoicacid, r-3-boc-amino-5-phenylpentanoic acid hplc, 3r-3-2-methylpropan-2-yl oxycarbonylamino-5-phenylpentanoic acid
PubChem CID 7021558
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-5-phenylpentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)CC(O)=O

Specifications

Quantity 1 g
Formula Weight 293.36
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-5-phenylpentanoic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.