Learn More
Medchemexpress LLC (S)-3-(Boc-amino)pyrrolidine | 122536-76-9 | 99.8% | C9H18N2O2 | 10 G
SDP
Catalog No. 5000432370
Encompass_Preferred
(S)-3-(Boc-amino)pyrrolidine is a biochemical reagent suitable for life science research and is also utilized as a pharmaceutical intermediate. It appears as a white to off-white solid.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Used as a pharmaceutical intermediate
  • Stores at 4°C, protected from light
  • In solvent, stores at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at ≥ 100 mg/mL
Catalog No. 50-004-32370 Supplier Medchemexpress LLC Supplier No. HY7969110G
(S)-3-(Boc-amino)pyrrolidine is a biochemical reagent suitable for life science research and is also utilized as a pharmaceutical intermediate. It appears as a white to off-white solid.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Used as a pharmaceutical intermediate
  • Stores at 4°C, protected from light
  • In solvent, stores at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at ≥ 100 mg/mL