Learn More
Medchemexpress LLC (S)-3-bromo-N-((1-ethyl-2-pyrrolidinyl)methyl)-2,6-dimethoxybenzamide hydrochloride | 73220-03-8 | >98.0% | 407.73 g/mol | C16H24BrClN2O3 | 5 MG
SDP
Catalog No. 5000212507
Encompass_Preferred
(S)-Remoxipride hydrochloride is the S-enantiomer of remoxipride supplied as the hydrochloride salt for use as a research reagent in dopamine D2 receptor pharmacology. It is supplied as a solid powder with reported high purity and is intended for in vitro and in vivo pharmacology studies, assay development, and analytical reference work. CAS: 73220-03-8.
  • S enantiomer composition suitable for stereospecific studies.
  • Hydrochloride salt provided as a solid powder for easy handling.
  • High purity suitable for research and analytical use.
  • Used in dopamine D2 receptor pharmacology and psychotropic disorder research.
  • Available in small research pack sizes for preclinical work.
Catalog No. 50-002-12507 Supplier Medchemexpress LLC Supplier No. HY101313A5MG
(S)-Remoxipride hydrochloride is the S-enantiomer of remoxipride supplied as the hydrochloride salt for use as a research reagent in dopamine D2 receptor pharmacology. It is supplied as a solid powder with reported high purity and is intended for in vitro and in vivo pharmacology studies, assay development, and analytical reference work. CAS: 73220-03-8.
  • S enantiomer composition suitable for stereospecific studies.
  • Hydrochloride salt provided as a solid powder for easy handling.
  • High purity suitable for research and analytical use.
  • Used in dopamine D2 receptor pharmacology and psychotropic disorder research.
  • Available in small research pack sizes for preclinical work.