Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-Chloro-1-phenyl-1-propanol 98.0+%, TCI America™
SDP

Catalog No. C24241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24241G Supplier TCI America Supplier No. C24241G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100306-34-1
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.636
MDL Number MFCD00013309
InChI Key JZFUHAGLMZWKTF-VIFPVBQESA-N
Synonym s---3-chloro-1-phenyl-1-propanol, s-3-chloro-1-phenylpropan-1-ol, s-3-chloro-1-phenyl-1-propanol, 1s-3-chloro-1-phenylpropan-1-ol, 1s-3-chloro-1-phenyl-propan-1-ol, s-3-chloro-1-phenylpropanol, s-3-chloro-1-phenyl-propan-1-ol, pubchem6275
PubChem CID 2777894
IUPAC Name (1S)-3-chloro-1-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCCl)O

Specifications

Melting Point 59°C
Color White
Quantity 1 g
Formula Weight 170.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-3-Chloro-1-phenyl-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.