Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-3-Fluoropyrrolidine hydrochloride | 136725-53-6 | 125.57 | 10 G
SDP

Catalog No. 5000446050
Encompass_Preferred

(S)-3-Fluoropyrrolidine hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. This product is intended for research use only.

  • Appears as a white to off-white solid
  • Purity: 99.95%
  • Molecular weight: 125.57
  • Chemical formula: C4H9ClFN
  • CAS number: 136725-53-6
  • Ships at room temperature (in continental US)
  • Store at 4°C, sealed, away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-46050 Supplier Medchemexpress LLC Supplier No. HYW00469510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-3-Fluoropyrrolidine hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. This product is intended for research use only.

  • Appears as a white to off-white solid
  • Purity: 99.95%
  • Molecular weight: 125.57
  • Chemical formula: C4H9ClFN
  • CAS number: 136725-53-6
  • Ships at room temperature (in continental US)
  • Store at 4°C, sealed, away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.