Learn More
Cayman Chemical S-3-hydroxy BlebbIstatIn 10mg
SDP
Catalog No. 502934610
Encompass_Preferred
A more stable form of (–)-blebbistatin; less inherent fluorescence than (–)-blebbistatin while retaining the activity; has the same stereochemistry as (–)-blebbistatin
Catalog No. 50-293-4610 Supplier Cayman Chemical Supplier No. 2416910 MG
A more stable form of (–)-blebbistatin; less inherent fluorescence than (–)-blebbistatin while retaining the activity; has the same stereochemistry as (–)-blebbistatin