Learn More
Medchemexpress LLC HY-W050031 50mg Medchemexpress, (S)-3-Hydroxybutanoic acid CAS:6168-83-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW05003150MG
Encompass_Preferred
Medchemexpress, HY-W050031 50mg (S)-3-Hydroxybutanoic acid CAS:6168-83-8 (S)-3-Hydroxybutanoic acid is a normal human metabolite, that has been found elevated in geriatric patients remitting from depression. In humans, 3-Hydroxybutyric acid is synthesized in the liver from acetyl-CoA, and can be used as an energy source by the brain when blood glucose is low. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4663