Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-Hydroxypyrrolidine, 97+%, ee 98+%

Catalog No. AAL1949803
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1949803 1 g
AAL1949806 5 g
AAL19498MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1949803 Supplier Thermo Scientific Chemicals Supplier No. L1949803
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 100243-39-8 | C4H10NO | 88.13 g/mol

(S)-(-)-3-Hydroxypyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Hydroxypyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is hygroscopic and sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100243-39-8
Molecular Formula C4H10NO
Molecular Weight (g/mol) 88.13
MDL Number MFCD00192426
InChI Key JHHZLHWJQPUNKB-BYPYZUCNSA-O
Synonym s-pyrrolidin-3-ol, s-3-hydroxypyrrolidine, 3s-pyrrolidin-3-ol, s-3-pyrrolidinol, s---3-pyrrolidinol, 3-pyrrolidinol, 3s, 3s-3-pyrrolidinol, 3s-3-hydroxypyrrolidine, s-3-hydroxy-pyrrolidine, s---3-hydroxypyrrolidine
PubChem CID 2733874
SMILES O[C@H]1CC[NH2+]C1

Specifications

Melting Point 15°C
Density 1.048
Boiling Point 108°C to 110°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.49
Quantity 1 g
Beilstein 4652606
Sensitivity Air sensitive; Hygroscopic
Solubility Information Fully miscible in water.
Optical Rotation −6.5° (c=3.5 in Methanol)
IUPAC Name (3S)-pyrrolidin-3-ol
Formula Weight 87.12
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (S)-(-)-3-Hydroxypyrrolidine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.