Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-3-methyl-pentanoic acid

Catalog No. CDS005352 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005352 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005352 Supplier Sigma Aldrich Supplier No. CDS005352100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-bromooctanoic acid is an effective substitute for the expensive (2,000 times) and cell-growth-inhibiting polyhydroxyalkanoic acid synthesis inhibitor, cerulenin
  • It blocks the formation of polyhydroxyalkanoic acid in pseudomonas fluorescens bm07 without any influence on the cell growth when grown on fructose
  • It inhibits α-oxidation of fatty acids in perfused rat liver and in mitochondria isolated from rat liver

Specifications

Molecular Formula C6H12O2
Linear Formula C6H12O2
MDL Number MFCD02262177
Quantity 10 g
Molecular Weight (g/mol) 116.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.