Learn More
Medchemexpress LLC S -3-N-Cbz-Amino-su 25g
SDP
Catalog No. 5000204721
Encompass_Preferred
(S)-3-N-Cbz-Amino-succinimide (Compound 1d) is an antiepileptic agent that can inhibit pentylenetetrazole (PTZ) and maximal electroshock (MES)-induced tonic convulsions in mice[1]
Catalog No. 50-002-04721 Supplier Medchemexpress LLC Supplier No. 25GHYW097106
(S)-3-N-Cbz-Amino-succinimide (Compound 1d) is an antiepileptic agent that can inhibit pentylenetetrazole (PTZ) and maximal electroshock (MES)-induced tonic convulsions in mice[1]