Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ (S)-3-(N-Fmoc-L-leucinyl)-2,2-dimethyloxazolidine-4-carboxylic acid, 95%

Catalog No. AAH66629MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66629MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66629MD Supplier Thermo Scientific™ Supplier No. H66629MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Purity 95%
CAS 339531-50-9
Content And Storage Ambient temperatures
Format Solid
Molecular Weight (g/mol) 480.561
Product Type (S)-3-(N-Fmoc-L-leucinyl)-2,2-dimethyloxazolidine-4-carboxylic acid
Quantity 250mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.