Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Phenylcyclopentanone 98.0+%, TCI America™
SDP

Catalog No. P1509100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1509100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1509100MG Supplier TCI America Supplier No. P1509100MG

missing translation for 'validMainframeMsg'

Specifications

CAS 86505-50-2
Color Yellow
Boiling Point 155°C
Molecular Formula C11H12O
Quantity 100 mg
InChI Key MFBXYJLOYZMFIN-JTQLQIEISA-N
SMILES C1CC(=O)CC1C2=CC=CC=C2
IUPAC Name (3S)-3-phenylcyclopentan-1-one
Molecular Weight (g/mol) 160.216
PubChem CID 10931730
Formula Weight 160.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-3-Phenylcyclopentanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.