Learn More
Cayman Chemical (S)-3-Thienylglycine, Purity: ≥95%, CAS: 1194-87-2, 100 mg
SDP
Catalog No. 502988461
Encompass_Preferred
A thienyl-containing amino acid; has been used as an affinity ligand to adsorb IgM and IgG antibodies from isolated human plasma
Catalog No. 50-298-8461 Supplier Cayman Chemical Supplier No. 30912100 MG
A thienyl-containing amino acid; has been used as an affinity ligand to adsorb IgM and IgG antibodies from isolated human plasma