Learn More
TARGETMOL CHEMICALS INC S 3304 5MG
SDP
Catalog No. 502581134
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. S 3304 is a novel matrix metalloproteinases inhibitor which is specific for MMP-2 and MMP-9. purity: 96%
Catalog No. 50-258-1134 Supplier TARGETMOL CHEMICALS INC Supplier No. T169665MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. S 3304 is a novel matrix metalloproteinases inhibitor which is specific for MMP-2 and MMP-9. purity: 96%