Learn More
Medchemexpress LLC HY-104003 5mg Medchemexpress, S 38093 CAS:862896-30-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1040035MG
Encompass_Preferred
Medchemexpress, HY-104003 5mg S 38093 CAS:862896-30-8 S 38093 is a brain-penetrant antagonist of H3 receptor, with Ki of 8.8, 1.44 and 1.2 µM for rat, mouse and human H3 receptors, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8181