Learn More
Selleck Chemical LLC S 38093
SDP
Catalog No. 5000002857
Encompass_Preferred
S 38093 is a histamine H3 antagonist/inverse agonist with a moderate affinity for rat mouse and human H3 receptors (Ki 8 8 1 44 and 1 2 M respectively) and no affinity for other histaminergic receptors
Catalog No. 50-000-02857 Supplier Selleck Chemical LLC Supplier No. S85982MG
S 38093 is a histamine H3 antagonist/inverse agonist with a moderate affinity for rat mouse and human H3 receptors (Ki 8 8 1 44 and 1 2 M respectively) and no affinity for other histaminergic receptors