Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-4-(2-(2-bromoacetamido)acetamido)-5-((3-(4-((6aR,6bS,7S,8aS,8bS)-7-hydroxy | 2734878-02-3 | 99.6% | 866.79 | C43H52BrN3O11 | 1 MG
SDP

Catalog No. 5000203028
Encompass_Preferred

INX-P is a glucocorticosteroid-target antibody drug conjugate (ADC) designed for research use. The compound targets the glucocorticoid receptor and is supplied as a solid with high reported purity and defined physicochemical properties.

  • Intended use: research use only, not for human therapeutic use.
  • Target: glucocorticoid receptor.
  • Form: solid, white to off-white.
  • Purity: 99.63% reported by certificate of analysis.
  • Molecular weight: 866.79.
  • Chemical formula: C43H52BrN3O11.
  • CAS number: 2734878-02-3.
  • Available pack sizes: 1 MG, 5 MG, 10 MG, larger sizes available by quote.

Catalog No. 50-002-03028 Supplier Medchemexpress LLC Supplier No. HY1601821MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

INX-P is a glucocorticosteroid-target antibody drug conjugate (ADC) designed for research use. The compound targets the glucocorticoid receptor and is supplied as a solid with high reported purity and defined physicochemical properties.

  • Intended use: research use only, not for human therapeutic use.
  • Target: glucocorticoid receptor.
  • Form: solid, white to off-white.
  • Purity: 99.63% reported by certificate of analysis.
  • Molecular weight: 866.79.
  • Chemical formula: C43H52BrN3O11.
  • CAS number: 2734878-02-3.
  • Available pack sizes: 1 MG, 5 MG, 10 MG, larger sizes available by quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.