Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-4-Amino-2-hydroxybutyric acid, 98%

Catalog No. AAH2879614
Click to view available options
Quantity:
5 g
25 g

CAS: 40371-51-5 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40371-51-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00674110
InChI Key IVUOMFWNDGNLBJ-VKHMYHEASA-N
Synonym s-4-amino-2-hydroxybutanoic acid, s-4-amino-2-hydroxybutyric acid, s---4-amino-2-hydroxybutyric acid, 2s-4-amino-2-hydroxybutanoic acid, s---amino-2-hydroxybutyric acid, butanoic acid, 4-amino-2-hydroxy-, 2s, s---4-amino-2-hydroxybutyricacid, pubchem23215, l-2-hydroxy-4-aminobutyric acid
PubChem CID 2733929
IUPAC Name (2S)-4-amino-2-hydroxybutanoic acid
SMILES [NH3+]CC[C@H](O)C([O-])=O

Specifications

Quantity 25 g
Formula Weight 119.12
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material (S)-4-Amino-2-hydroxybutyric acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.