Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-4-amino-3-hydroxybutanoic acid | 7013-05-0 | MFCD00672884 | 95.0% | 119.12 g·mol⁻1 | C4H9NO3 | 25 G
SDP

Catalog No. 5000208847
Encompass_Preferred

S-(+)-GABOB (S-4-amino-3-hydroxybutanoic acid) is the S-enantiomer of GABOB, an endogenous GABA metabolite with reported antiepileptic activity. Supplied as a research-grade reagent with 95.0% purity, it is intended for laboratory research use only.

  • 95.0% purity
  • Molecular weight 119.12 g·mol⁻1
  • Chemical formula C4H9NO3
  • SMILES: O=C(C[C@@H](CN)O)O
  • Available as a 25 G pack for laboratory use
  • For research use only; not for human or clinical applications

Catalog No. 50-002-08847 Supplier Medchemexpress LLC Supplier No. 25GHYW050122
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-(+)-GABOB (S-4-amino-3-hydroxybutanoic acid) is the S-enantiomer of GABOB, an endogenous GABA metabolite with reported antiepileptic activity. Supplied as a research-grade reagent with 95.0% purity, it is intended for laboratory research use only.

  • 95.0% purity
  • Molecular weight 119.12 g·mol⁻1
  • Chemical formula C4H9NO3
  • SMILES: O=C(C[C@@H](CN)O)O
  • Available as a 25 G pack for laboratory use
  • For research use only; not for human or clinical applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.