Learn More
Medchemexpress LLC (S)-4-Azido-2-hydroxybutanoate dicyclohexylamine | C16H30N4O3 | 25 MG
SDP
Catalog No. 5000374395
Encompass_Preferred
(S)-N3-HABA (DCHA) is a click chemistry reagent designed for high-efficiency binding between nucleic acids, lipids, proteins, and other molecules. This reagent is highly valued in various research fields due to its high yield, high specificity, and simplicity in reactions. It features an azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) and ring strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Specifically designed for click chemistry applications.
  • Characterized by high yield, high specificity, and simplicity in reactions.
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) and ring strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Useful for binding nucleic acids, lipids, proteins, and other molecules in diverse research applications.
Catalog No. 50-003-74395 Supplier Medchemexpress LLC Supplier No. 25MGHY151866A
(S)-N3-HABA (DCHA) is a click chemistry reagent designed for high-efficiency binding between nucleic acids, lipids, proteins, and other molecules. This reagent is highly valued in various research fields due to its high yield, high specificity, and simplicity in reactions. It features an azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) and ring strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Specifically designed for click chemistry applications.
  • Characterized by high yield, high specificity, and simplicity in reactions.
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) and ring strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Useful for binding nucleic acids, lipids, proteins, and other molecules in diverse research applications.