Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-4-Chloro-Alpha-Methylbenzyl Alcohol

Catalog No. 6862711G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-627-11G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 68-627-11G Supplier Sigma Aldrich Supplier No. 6862711G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-4-Chloro-+a-methylbenzyl alcohol can be used as a chiral building block in: The enantioselective synthesis of (-)-(S,S)-clemastine. The enantioselective and geometrically defined synthesis of chloro methylbenzyl pinacol boronic ester via lithiation-borylation methodology.

Specifications

CAS 210169-54-3
Melting Point 231°C to 235°C
Molecular Formula C38H28O4P2
Refractive Index n20/D 1.544
Linear Formula C38H28O4P2
MDL Number MFCD09753005
Quantity 100 mg
Synonym (S)-1-(4-Chlorophenyl)ethanol; (S)-4'-Chloro-1-phenylethanol
Molecular Weight (g/mol) 610.57 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.