Learn More
Strem, An Ascensus Company CAS# 137156-22-0. 5g. (S)-(+)-4-Chlorodinaphthol[2,1-d:1',2'-f][1,3,2]dioxaphosphepin, min. 97%. MFCD02093048
SDP
Supplier:  Strem, An Ascensus Company 1506865G
Encompass
CAS# 137156-22-0. 5g. (S)-(+)-4-Chlorodinaphthol[2,1-d:1',2'-f][1,3,2]dioxaphosphepin, min. 97%. MFCD02093048. Molecular Weight: 350.74. Molecular Formula: C20H12ClO2P. Color/form: white pwdr. Strem# 15-0686. http://www.strem.com/catalog/v/15-0686/
Catalog No. 50-901-09964