Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-(4-Hydroxybenzyl)glutathione | 129636-38-0 | 5 MG
SDP

Catalog No. 5000418240
Encompass_Preferred

S-(4-Hydroxybenzyl)glutathione is a glutathione derivative that inhibits the in vitro binding of kainic acid to brain glutamate receptors, with an IC50 of 2 μM. It is intended for research use only.

  • Glutathione derivative
  • Inhibits kainic acid binding to brain glutamate receptors
  • IC50 of 2 μM
  • Molecular weight: 413.45
  • Molecular formula: C17H23N3O7S
  • Appearance: White to off-white solid
  • Isolated from plants, Orchidaceae, Gastrodia elata Bl.

Catalog No. 50-004-18240 Supplier Medchemexpress LLC Supplier No. HYN81155MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-(4-Hydroxybenzyl)glutathione is a glutathione derivative that inhibits the in vitro binding of kainic acid to brain glutamate receptors, with an IC50 of 2 μM. It is intended for research use only.

  • Glutathione derivative
  • Inhibits kainic acid binding to brain glutamate receptors
  • IC50 of 2 μM
  • Molecular weight: 413.45
  • Molecular formula: C17H23N3O7S
  • Appearance: White to off-white solid
  • Isolated from plants, Orchidaceae, Gastrodia elata Bl.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.