Learn More
Medchemexpress LLC (S)-4-isopropyl-3-methyl-2-(3-methylpiperidine-1-carbonyl)isoxazol-5(2H)-one | 654059-21-9 | 99.8% | 266.34 g/mol | C14H22N2O3 | 100 MG
SDP
Catalog No. 5000248334
Encompass_Preferred
BAY 59-9435 (HSL-IN-2) is a potent, selective small-molecule inhibitor of hormone-sensitive lipase (HSL) with an IC50 of 0.023 μM. It exhibits cellular activity (28 nM in 3T3-L1 cells for inhibition of forskolin-stimulated lipolysis) and is supplied as a high-purity solid for research use, with recommended storage conditions to maintain stability.
  • Potent and selective HSL inhibition (IC50 0.023 μM).
  • Demonstrated cellular activity in adipocyte assays (28 nM).
  • High purity suitable for biochemical and cellular studies.
  • Stable as powder under recommended storage conditions.
  • Soluble in DMSO for assay preparation.
Catalog No. 50-002-48334 Supplier Medchemexpress LLC Supplier No. HY102056100MG
BAY 59-9435 (HSL-IN-2) is a potent, selective small-molecule inhibitor of hormone-sensitive lipase (HSL) with an IC50 of 0.023 μM. It exhibits cellular activity (28 nM in 3T3-L1 cells for inhibition of forskolin-stimulated lipolysis) and is supplied as a high-purity solid for research use, with recommended storage conditions to maintain stability.
  • Potent and selective HSL inhibition (IC50 0.023 μM).
  • Demonstrated cellular activity in adipocyte assays (28 nM).
  • High purity suitable for biochemical and cellular studies.
  • Stable as powder under recommended storage conditions.
  • Soluble in DMSO for assay preparation.