Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-4-Methoxy-Alpha-Methylbenzylamine

Catalog No. 72665625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-665-625G 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 72-665-625G Supplier Sigma Aldrich Supplier No. 72665625G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-4-Methoxy-+a-methylbenzylamine is employed in the synthesis of S(+)-4-(1-phenylethylamino)quinazolines, as human immunoglobuline E inhibitor and haloaryl-+a-amino acids. It is also used as a precursor to prepare chiral intermediate in the total synthesis of solanoeclepin A.ChIpr∗

Specifications

CAS 86520-52-7
Molecular Formula C6H13N3O3
Linear Formula C6H13N3O3
MDL Number MFCD09753542
Quantity 10 mL
Synonym 8-Azido-3,6-dioxaoctanol solution
Molecular Weight (g/mol) 175.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.