Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-(4-Methoxybenzyl)-L-cysteine, tech. 80%, Thermo Scientific™

Catalog No. AAH6389414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6389414 25 g
AAH6389406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6389414 Supplier Thermo Scientific Chemicals Supplier No. H6389414

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2544-31-2
Molecular Formula C11H15NO3S
Molecular Weight (g/mol) 241.305
MDL Number MFCD00037207
InChI Key PQPZSPJVMUCVAQ-JTQLQIEISA-N
Synonym h-cys pmeobzl-oh, h-cys 4-meo-bzl-oh, s-4-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxybenzyl thio propanoic acid, s-4-methoxybenzyl cysteine, s-p-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxy-benzylsulfanyl-propionic acid, 2-amino-3-4-methoxybenzyl thio propanoic acid, 2r-2-amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, ambotzhaa6100
PubChem CID 7009563
IUPAC Name (2R)-2-amino-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid
SMILES COC1=CC=C(C=C1)CSCC(C(=O)O)N

Specifications

Quantity 25 g
Formula Weight 241.31
Percent Purity 80%
Grade Technical
Physical Form Solid
Chemical Name or Material S-(4-Methoxybenzyl)-L-cysteine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.