Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-4-Methyl-2-pentanol, 99%

Catalog No. AAL1888203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1888203 1 g
AAL18882MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1888203 Supplier Thermo Scientific Chemicals Supplier No. L1888203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14898-80-7 | C6H14O | 102.177 g/mol

It is employed in the reaction of 2-methoxyethyl acetals with allylsilanes in the presence of titanium tetrachloride: regioselective carbon-oxygen bond cleavage of the unsymmetrical acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the reaction of 2-methoxyethyl acetals with allylsilanes in the presence of titanium tetrachloride: regioselective carbon-oxygen bond cleavage of the unsymmetrical acetals.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-80-7
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD03093078
InChI Key WVYWICLMDOOCFB-LURJTMIESA-N
PubChem CID 6994471
IUPAC Name (2S)-4-methylpentan-2-ol
SMILES CC(C)CC(C)O

Specifications

Density 0.802
Boiling Point 130°C to 131°C
Flash Point 41°C (105°F)
Odor Slightly Sweetish
Refractive Index 1.411
Quantity 1 g
UN Number UN2053
Beilstein 1718991
Synonym s-+-4-methyl-2-pentanol, 2s-4-methylpentan-2-ol, s-+-4-methylpentan-2-ol, 4-methyl s-2-pentanol, s-4-methyl-pentan-2-ol, 2s-4-methyl-2-pentanol, s-1,3-dimethyl-1-butanol, 2-pentanol,4-methyl-, 2s, 2-pentanol, 4-methyl-, 2s, unii-8u34xjk0r0 component
Solubility Information Slightly soluble in water. Soluble in alcohol.
Optical Rotation +21° (Neat)
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material (S)-(+)-4-Methyl-2-pentanol
Show More Show Less
Hazard Category H226-H335
Hazard Statement GHS H Statement
H226-H335
Flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYL ISOBUTYL CARBINOL
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.