Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-4-Penten-2-Ol

Catalog No. 5580285G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-802-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 55-802-85G Supplier Sigma Aldrich Supplier No. 5580285G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-4-Penten-2-ol can be used:As an intermediate in one of the key synthetic steps for the preparation of undecenolide (-)-cladospolide C.To prepare a carboxamide derivative as a key intermediate for the preparation of (-)-iso-cladospolide B1.To prepare S-enantiomers of hydroxyeicosa tetraenoic acid named 19-HETE.To prepare an acryloyl ester derivative by reacting with acryloyl chloride, which is a key intermediate for the synthesis of parasorbic acid.

Specifications

CAS 64584-92-5
Density 0.837 g/mL at 25°C
Boiling Point 115°C to 116°C
Molecular Formula C5H10O
Refractive Index n20/D 1.4240
Linear Formula H2C=CHCH2CH(OH)CH3
MDL Number MFCD03701536
Quantity 5 g
Molecular Weight (g/mol) 86.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.