Learn More
Chemglass Life Sciences S-40 Ball Joint, Compatible with Buchi Rotary Evaporators, Tube OD: 34mm
SDP
Catalog No. CG12301
Encompass_Preferred
Special S-40 size ball joint compatible with Buchi rotary evaporator components. Both the ball and socket are tooled on Duran 34 mm O.D. x 2.8mm wall tubing.
Catalog No. CG12301 Supplier Chemglass Life Sciences Supplier No. CG12301
Special S-40 size ball joint compatible with Buchi rotary evaporator components. Both the ball and socket are tooled on Duran 34 mm O.D. x 2.8mm wall tubing.