Learn More
Chemglass Life Sciences S-40 Socket Joint, Compatible with Buchi Rotary Evaporators, Tube OD: 34mm
SDP
Catalog No. CG12302
Encompass_Preferred
Special S-40 size socket compatible with Buchi rotary evaporator components. Both the ball and socket are tooled on Duran 34 mm O.D. x 2.8mm wall tubing.
Catalog No. CG12302 Supplier Chemglass Life Sciences Supplier No. CG12302
Special S-40 size socket compatible with Buchi rotary evaporator components. Both the ball and socket are tooled on Duran 34 mm O.D. x 2.8mm wall tubing.