Learn More
Strem, An Ascensus Company CAS# 185913-98-8. (S)-(-)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl, min. 95% (S)-Cl-MeO-BIPHEP. MFCD03426987. 250MG
SDP
Catalog No. 5090110058
Encompass
CAS# 185913-98-8. 250MG. (S)-(-)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl, min. 95% (S)-Cl-MeO-BIPHEP MFCD03426987. Molecular Formula: C38H30Cl2O2P2. Molecular Weight: 651.50. Color/Form: yellow-white pwdr. Strem#: 15-1056 www.strem.com/catalog/v3/15-1056/
Catalog No. 50-901-10058 Supplier Strem, An Ascensus Company Supplier No. 151056250MG
CAS# 185913-98-8. 250MG. (S)-(-)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl, min. 95% (S)-Cl-MeO-BIPHEP MFCD03426987. Molecular Formula: C38H30Cl2O2P2. Molecular Weight: 651.50. Color/Form: yellow-white pwdr. Strem#: 15-1056 www.strem.com/catalog/v3/15-1056/