Learn More
AdipoGen S-5-Adenosyl-3-thiopropylam
SDP
Supplier:  AdipoGen CDXA0210M010
Encompass_Preferred
Chemical. CAS 53186-57-5. Formula C13H20N6O3S. MW 340.4. Synthetic This decarboxylated S-Adenosyl-L-methionine is an important metabolite in polyamine biosynthesis, acting as aminopropyl group donor for propylamine transferases such as spermine synthase and spermidine synthase.
Catalog No. 50-256-1794