Learn More
Cayman Chemical S-(5'-Adenosyl)-L-methionine (tosylate), Purity: ≥90%, CAS: 52248-03-0, 50 mg
SDP
Catalog No. 502972037
Encompass_Preferred
A methyl donor and cofactor; involved in several biological processes, including epigenetic regulation, post-translational modification of proteins, and metabolism
Catalog No. 50-297-2037 Supplier Cayman Chemical Supplier No. 1637650 MG
A methyl donor and cofactor; involved in several biological processes, including epigenetic regulation, post-translational modification of proteins, and metabolism