Learn More
TARGETMOL CHEMICALS INC S-5-Adenosyl-L-methionine tosy
SDP
Catalog No. 502878707
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. S-(5'-Adenosyl)-L-methionine tosylate (SAMe) is a ubiquitous methyl donor involved in a wide variety of biological reactions, including those mediated by DNA and protein methyltransferases [1]. S-(5'-Adenosyl)-L-methionine tosylate (SAMe) is a stable salt of SAM that is included in nutritional supplements for oral use [2]. S-(5'-Adenosyl)-L-methionine tosylate (SAMe) reportedly ameliorates depression, pain associated with osteoarthritis and fibromyalgia, and liver toxicity [3]. Purity 85.51%
Catalog No. 50-287-8707 Supplier TARGETMOL CHEMICALS INC Supplier No. T4447200MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. S-(5'-Adenosyl)-L-methionine tosylate (SAMe) is a ubiquitous methyl donor involved in a wide variety of biological reactions, including those mediated by DNA and protein methyltransferases [1]. S-(5'-Adenosyl)-L-methionine tosylate (SAMe) is a stable salt of SAM that is included in nutritional supplements for oral use [2]. S-(5'-Adenosyl)-L-methionine tosylate (SAMe) reportedly ameliorates depression, pain associated with osteoarthritis and fibromyalgia, and liver toxicity [3]. Purity 85.51%