Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene 98.0+%, TCI America™
SDP

Catalog No. A19841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19841G Supplier TCI America Supplier No. A19841G

Chemical Identifiers

CAS 1052236-86-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
InChI Key WBRYUDKSKFXYOD-SNVBAGLBSA-N
PubChem CID 57372770
IUPAC Name (3aS)-3a-prop-2-enyl-2,3,4,5-tetrahydrocyclopenta[b]furan
SMILES C=CCC12CCC=C1OCC2

Specifications

Color Yellow
Quantity 1 g
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.