Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-5-Benzyl-2-Mesityl-6,6-Dimethyl-6,8-Dihydro-5H-[1,2,4]Triazolo[3,4-C][1,4]Oxazin-2-Ium Tetrafluoroborate

Catalog No. 708569100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-856-9100MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-856-9100MG Supplier Sigma Aldrich Supplier No. 708569100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-5-Benzyl-2-Mesityl-6,6-Dimethyl-6,8-Dihydro-5H-[1,2,4]Triazolo[3,4-C][1,4]Oxazin-2-Ium Tetrafluoroborate

Specifications

Melting Point 183°C
Molecular Formula C3F9FeO9S3
Linear Formula C3F9FeO9S3
MDL Number MFCD15144785
Quantity 1 g
Synonym Fe(CF3SO3)3; Ferric triflate; Ferric trifluoromethanesulfonate; Fe(CF3SO3)3; Fe(OTf)3; Ferric triflate; Ferric trifluoromethanesulfonate; Iron 1,1,1-trifluoromethanesulfonic acid; Iron(III) triflate
Molecular Weight (g/mol) 503.05 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.